Product Details

SNP ID
rs148197321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29731312 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCAGCTCCTGAACTGGTGGCCA[G/T]CCCACGGGGTACTGGAAGACAGTGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CABP7 PubMed Links

Gene Details

Gene
CABP7
Gene Name
calcium binding protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182527.2 2461 UTR 3 NP_872333.1
XM_006724161.3 2461 Intron XP_006724224.1
Gene
ZMAT5
Gene Name
zinc finger matrin-type 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003692.1 2461 Silent Mutation GGA,GGC G142G NP_001003692.1
NM_001318129.1 2461 Silent Mutation GGA,GGC G142G NP_001305058.1
NM_019103.2 2461 Silent Mutation GGA,GGC G142G NP_061976.1

View Full Product Details