Product Details

SNP ID
rs148271273
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:44495457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTCATTTGGCATTGAAATATGG[C/T]GGAACAGCACAGTCCAAAGGGCGTG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LDOC1L PubMed Links

Gene Details

Gene
LDOC1L
Gene Name
leucine zipper down-regulated in cancer 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032287.2 2437 UTR 3 NP_115663.2

View Full Product Details