Product Details

SNP ID
rs148339904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:22245208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAACAGGGGGTATTTGAGCATCTC[C/T]GAGCTGCAGCCTGAGGACGAGGCTA
Phenotype
MIM: 605141
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VPREB1 PubMed Links

Gene Details

Gene
VPREB1
Gene Name
pre-B lymphocyte 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303509.1 329 Silent Mutation TCC,TCT S102S NP_001290438.1
NM_007128.3 329 Silent Mutation TCC,TCT S103S NP_009059.1

View Full Product Details