Product Details

SNP ID
rs149095723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40773952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGCTCACCTTACTCGTTGGTGA[A/G]GAAGATAGAGAATATTCCGAAGACT
Phenotype
MIM: 606795
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A17 PubMed Links

Gene Details

Gene
SLC25A17
Gene Name
solute carrier family 25 member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282726.1 965 Missense Mutation CCT,CTT P217L NP_001269655.1
NM_001282727.1 965 Missense Mutation CCT,CTT P181L NP_001269656.1
NM_006358.3 965 Missense Mutation CCT,CTT P254L NP_006349.1

View Full Product Details