Product Details
- SNP ID
-
rs149993265
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:43530867 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGTTCCAGCCTCATCATAGCTT[C/T]TGAACGTGCGCCGCATTGGCCTCCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EFCAB6
PubMed Links
Gene Details
- Gene
- EFCAB6
- Gene Name
- EF-hand calcium binding domain 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022785.3 |
4343 |
Missense Mutation |
AAA,AGA |
K1444R |
NP_073622.2 |
NM_198856.2 |
4343 |
Missense Mutation |
AAA,AGA |
K1292R |
NP_942153.1 |
XM_005261704.2 |
4343 |
Missense Mutation |
AAA,AGA |
K1444R |
XP_005261761.1 |
XM_005261705.3 |
4343 |
Missense Mutation |
AAA,AGA |
K1338R |
XP_005261762.1 |
XM_011530316.1 |
4343 |
Missense Mutation |
AAA,AGA |
K1470R |
XP_011528618.1 |
XM_011530317.2 |
4343 |
Missense Mutation |
AAA,AGA |
K1444R |
XP_011528619.1 |
XM_011530318.2 |
4343 |
Missense Mutation |
AAA,AGA |
K1418R |
XP_011528620.1 |
XM_011530319.2 |
4343 |
Missense Mutation |
AAA,AGA |
K1338R |
XP_011528621.1 |
XM_011530320.2 |
4343 |
Missense Mutation |
AAA,GAA |
K1469E |
XP_011528622.1 |
XM_011530321.2 |
4343 |
Missense Mutation |
AAA,AGA |
K1292R |
XP_011528623.1 |
XM_011530322.2 |
4343 |
Missense Mutation |
AAA,AGA |
K1292R |
XP_011528624.1 |
XM_011530323.2 |
4343 |
Missense Mutation |
AAA,AGA |
K1290R |
XP_011528625.1 |
XM_011530325.2 |
4343 |
Missense Mutation |
AAA,AGA |
K1196R |
XP_011528627.1 |
XM_011530326.2 |
4343 |
Intron |
|
|
XP_011528628.1 |
XM_011530327.2 |
4343 |
Missense Mutation |
AAA,AGA |
K1136R |
XP_011528629.1 |
XM_011530328.2 |
4343 |
Intron |
|
|
XP_011528630.1 |
XM_011530329.1 |
4343 |
Missense Mutation |
AAA,AGA |
K742R |
XP_011528631.1 |
XM_011530330.2 |
4343 |
Intron |
|
|
XP_011528632.1 |
XM_017028910.1 |
4343 |
Missense Mutation |
AAA,AGA |
K1470R |
XP_016884399.1 |
XM_017028911.1 |
4343 |
Missense Mutation |
AAA,AGA |
K1470R |
XP_016884400.1 |
XM_017028912.1 |
4343 |
Missense Mutation |
AAA,AGA |
K1286R |
XP_016884401.1 |
- Gene
- EFCAB6-AS1
- Gene Name
- EFCAB6 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details