Product Details
- SNP ID
-
rs150583153
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:41464850 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGTAATTAACAAACACCTACTAT[A/G]TGCTAGACACCCTACAGGGGATACC
- Phenotype
-
MIM: 100850
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACO2
PubMed Links
Gene Details
- Gene
- ACO2
- Gene Name
- aconitase 2
There are no transcripts associated with this gene.
- Gene
- PHF5A
- Gene Name
- PHD finger protein 5A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032758.3 |
|
Intron |
|
|
NP_116147.1 |
View Full Product Details