Product Details

SNP ID
rs138273341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96853422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCTTATGCAGACTGGTCATGCC[A/G]TCGTCATCCACCACCCTGGGGTTGG
Phenotype
MIM: 610736 MIM: 604462
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD23 PubMed Links

Gene Details

Gene
ANKRD23
Gene Name
ankyrin repeat domain 23
There are no transcripts associated with this gene.

Gene
ANKRD39
Gene Name
ankyrin repeat domain 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016466.5 419 Silent Mutation GAC,GAT D129D NP_057550.3
Gene
SEMA4C
Gene Name
semaphorin 4C
There are no transcripts associated with this gene.

View Full Product Details