Product Details

SNP ID
rs138397308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:202880874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGTTCACTTTCATGCCCCAACAT[A/G]GGAAGTGGTAGGTGAATATCTGTAG
Phenotype
MIM: 616620
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ICA1L PubMed Links

Gene Details

Gene
ICA1L
Gene Name
islet cell autoantigen 1 like
There are no transcripts associated with this gene.

Gene
WDR12
Gene Name
WD repeat domain 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018256.3 1885 Missense Mutation CAT,TAT H420Y NP_060726.3
XM_011511469.2 1885 Missense Mutation CAT,TAT H282Y XP_011509771.1

View Full Product Details