Product Details

SNP ID
rs139281019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:108294262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATTCAGGAAATTGTGGATATGA[C/T]TGAACAGAATGGGGACGTGGAGAAG
Phenotype
MIM: 602385
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SULT1C2 PubMed Links

Gene Details

Gene
SULT1C2
Gene Name
sulfotransferase family 1C member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001056.3 638 Missense Mutation ACT,ATT T62I NP_001047.1
NM_176825.2 638 Missense Mutation ACT,ATT T62I NP_789795.1
XM_005264013.3 638 Missense Mutation ACT,ATT T64I XP_005264070.1

View Full Product Details