Product Details
- SNP ID
-
rs139679723
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:38984272 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCACAGACATTATAGTGGAGATTA[A/T]TAAAATGTTTTCTCTTCAACAAGGT
- Phenotype
-
MIM: 182530
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGEF33
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs34248802] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARHGEF33
- Gene Name
- Rho guanine nucleotide exchange factor 33
There are no transcripts associated with this gene.
- Gene
- SOS1
- Gene Name
- SOS Ras/Rac guanine nucleotide exchange factor 1
View Full Product Details