Product Details

SNP ID
rs139682365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:32633987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGGACTTTTTGTCATCTGTTTT[C/G]TTCCAGCAATTCAGTGAGGTATGCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR4765 PubMed Links

Gene Details

Gene
MIR4765
Gene Name
microRNA 4765
There are no transcripts associated with this gene.

Gene
TTC27
Gene Name
tetratricopeptide repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193509.1 651 Silent Mutation TTC,TTG F76L NP_001180438.1
NM_017735.4 651 Silent Mutation TTC,TTG F126L NP_060205.3
XM_005264416.1 651 Missense Mutation TTC,TTG F126L XP_005264473.1
XM_011532958.1 651 Missense Mutation TTC,TTG F126L XP_011531260.1

View Full Product Details