Product Details

SNP ID
rs139801918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:61190366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTTATCAAGTTTGTAATAAGAG[A/T]GCTGATCAAATTGGCACAGTTTGCT
Phenotype
MIM: 615295
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AHSA2 PubMed Links

Gene Details

Gene
AHSA2
Gene Name
AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321300.1 3616 Intron NP_001308229.1
NM_001321301.1 3616 Intron NP_001308230.1
NM_001321302.1 3616 Intron NP_001308231.1
NM_001321303.1 3616 Intron NP_001308232.1
NM_001321304.1 3616 Intron NP_001308233.1
NM_001321305.1 3616 Intron NP_001308234.1
NM_152392.4 3616 Intron NP_689605.1
XM_011532541.2 3616 UTR 3 XP_011530843.1
XM_011532543.2 3616 UTR 3 XP_011530845.1
XM_011532545.2 3616 UTR 3 XP_011530847.1
XM_017003377.1 3616 UTR 3 XP_016858866.1
XM_017003378.1 3616 UTR 3 XP_016858867.1
Gene
USP34
Gene Name
ubiquitin specific peptidase 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014709.3 3616 Missense Mutation ACT,TCT T3260S NP_055524.3

View Full Product Details