Product Details

SNP ID
rs140081455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:3645459 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCGGGCTGTTCTGCATAATCAAT[C/T]AGCTTAGGGAACTAGCAGAGTGAGA
Phenotype
MIM: 612396 MIM: 612502
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALLC PubMed Links

Gene Details

Gene
ALLC
Gene Name
allantoicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018436.3 92 Intron NP_060906.3
XM_011510369.2 92 Intron XP_011508671.1
XM_011510370.2 92 Intron XP_011508672.1
XM_017004495.1 92 UTR 5 XP_016859984.1
XM_017004496.1 92 Intron XP_016859985.1
XM_017004497.1 92 Intron XP_016859986.1
XM_017004498.1 92 Intron XP_016859987.1
Gene
COLEC11
Gene Name
collectin subfamily member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001255982.1 92 Intron NP_001242911.1
NM_001255983.1 92 Intron NP_001242912.1
NM_001255984.1 92 Intron NP_001242913.1
NM_001255985.1 92 Intron NP_001242914.1
NM_001255986.1 92 Intron NP_001242915.1
NM_001255987.1 92 Intron NP_001242916.1
NM_001255988.1 92 Intron NP_001242917.1
NM_001255989.1 92 Intron NP_001242918.1
NM_024027.4 92 Intron NP_076932.1
NM_199235.2 92 Intron NP_954705.1
XM_005263853.4 92 Intron XP_005263910.1
XM_006711897.3 92 Intron XP_006711960.1

View Full Product Details