Product Details
- SNP ID
-
rs140107070
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:96861007 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGCTCCTTGGGCAGCTCCAGGGG[A/G]TACACCAAGGTCCTCTCAGTAGCCT
- Phenotype
-
MIM: 604462
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD39
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12471298,rs12476284] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANKRD39
- Gene Name
- ankyrin repeat domain 39
There are no transcripts associated with this gene.
- Gene
- SEMA4C
- Gene Name
- semaphorin 4C
View Full Product Details