Product Details

SNP ID
rs140566168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:177625904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTGTCCACTGCTTCATAGTCA[C/T]GGAGACACAGACCCATGGCATTAGT
Phenotype
MIM: 604961
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDE11A PubMed Links

Gene Details

Gene
PDE11A
Gene Name
phosphodiesterase 11A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077196.1 5124 UTR 3 NP_001070664.1
NM_001077197.1 5124 UTR 3 NP_001070665.1
NM_001077358.1 5124 Intron NP_001070826.1
NM_016953.3 5124 UTR 3 NP_058649.3
Gene
TTC30A
Gene Name
tetratricopeptide repeat domain 30A
There are no transcripts associated with this gene.

View Full Product Details