Product Details

SNP ID
rs140791647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24038523 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCGTCATGGTAACTGCCTTCTT[A/G]AAGGTCACAAGGACCAAATGTGAAG
Phenotype
MIM: 600620 MIM: 616234
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FKBP1B PubMed Links

Gene Details

Gene
FKBP1B
Gene Name
FK506 binding protein 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322963.1 1103 Intron NP_001309892.1
NM_001322964.1 1103 Intron NP_001309893.1
NM_004116.4 1103 Intron NP_004107.1
NM_054033.3 1103 Intron NP_473374.1
XM_011532699.2 1103 Intron XP_011531001.1
XM_017003593.1 1103 Intron XP_016859082.1
XM_017003594.1 1103 Intron XP_016859083.1
XM_017003595.1 1103 Intron XP_016859084.1
XM_017003596.1 1103 Intron XP_016859085.1
Gene
WDCP
Gene Name
WD repeat and coiled coil containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142319.1 1103 Silent Mutation TTC,TTT F324F NP_001135791.1
NM_025203.2 1103 Silent Mutation TTC,TTT F324F NP_079479.1
XM_017005029.1 1103 Silent Mutation TTC,TTT F324F XP_016860518.1

View Full Product Details