Product Details

SNP ID
rs140904588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:11679202 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCATCGCTATTCCCATTTGACA[A/G]CTGAGAAAACTGAGGCTATGAAGTT
Phenotype
MIM: 605518 MIM: 605538
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100506405 PubMed Links

Gene Details

Gene
LOC100506405
Gene Name
uncharacterized LOC100506405
There are no transcripts associated with this gene.

Gene
LPIN1
Gene Name
lipin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261427.1 Intron NP_001248356.1
NM_001261428.1 Intron NP_001248357.1
NM_001261429.1 Intron NP_001248358.1
NM_145693.2 Intron NP_663731.1
XM_006711869.2 Intron XP_006711932.1
XM_006711870.3 Intron XP_006711933.1
XM_006711871.2 Intron XP_006711934.1
XM_006711872.2 Intron XP_006711935.1
XM_006711874.2 Intron XP_006711937.1
XM_011510333.2 Intron XP_011508635.1
XM_011510334.2 Intron XP_011508636.1
XM_011510335.2 Intron XP_011508637.1
XM_011510336.2 Intron XP_011508638.1
XM_017003623.1 Intron XP_016859112.1
XM_017003624.1 Intron XP_016859113.1
XM_017003625.1 Intron XP_016859114.1
XM_017003626.1 Intron XP_016859115.1
XM_017003627.1 Intron XP_016859116.1
XM_017003628.1 Intron XP_016859117.1
XM_017003629.1 Intron XP_016859118.1
XM_017003630.1 Intron XP_016859119.1
XM_017003631.1 Intron XP_016859120.1
XM_017003632.1 Intron XP_016859121.1
Gene
NTSR2
Gene Name
neurotensin receptor 2
There are no transcripts associated with this gene.

View Full Product Details