Product Details

SNP ID
rs141388155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:98799962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTCCTGACCCTCAGCCGTCATC[A/G]CTTCCTCCCAGCATTCTCTCTTCTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA1211L PubMed Links

Gene Details

Gene
KIAA1211L
Gene Name
KIAA1211 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207362.2 Intron NP_997245.2
XM_005263923.3 Intron XP_005263980.1
XM_005263924.3 Intron XP_005263981.1
XM_005263926.3 Intron XP_005263983.1
XM_011511093.1 Intron XP_011509395.1
XM_011511094.1 Intron XP_011509396.1
XM_011511095.2 Intron XP_011509397.1
XM_011511096.1 Intron XP_011509398.1
XM_017003971.1 Intron XP_016859460.1
XM_017003972.1 Intron XP_016859461.1
XM_017003973.1 Intron XP_016859462.1

View Full Product Details