Product Details

SNP ID
rs141846981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:150470149 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTACACATGCCAAGGTGGCAAC[A/G]TGAAAAATGTCTCTGACGCTATTTT
Phenotype
MIM: 602924
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RND3 PubMed Links

Gene Details

Gene
RND3
Gene Name
Rho family GTPase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001254738.1 851 Silent Mutation CAC,CAT H191H NP_001241667.1
NM_005168.4 851 Silent Mutation CAC,CAT H191H NP_005159.1

View Full Product Details