Product Details

SNP ID
rs141925489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:53795951 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTAGGAAGAAGAAAAGGATTAT[A/C]AAGGCCCTAATCCAAGAGAGCTTTT
Phenotype
MIM: 605760 MIM: 611229
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ASB3 PubMed Links

Gene Details

Gene
ASB3
Gene Name
ankyrin repeat and SOCS box containing 3
There are no transcripts associated with this gene.

Gene
ERLEC1
Gene Name
endoplasmic reticulum lectin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127397.2 566 Missense Mutation AAA,CAA K96Q NP_001120869.1
NM_001127398.2 566 Missense Mutation AAA,CAA K96Q NP_001120870.1
NM_015701.4 566 Missense Mutation AAA,CAA K96Q NP_056516.2
XM_011532766.1 566 Missense Mutation AAA,CAA K96Q XP_011531068.1
Gene
GPR75-ASB3
Gene Name
GPR75-ASB3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164165.1 566 Intron NP_001157637.1

View Full Product Details