Product Details

SNP ID
rs142350544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219237133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGTATCAGCTGAAAGGGAATTG[A/G]TATGTGTCCTGTGCAAAGTACTAGT
Phenotype
MIM: 612204 MIM: 604719
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKZF1 PubMed Links

Gene Details

Gene
ANKZF1
Gene Name
ankyrin repeat and zinc finger domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042410.1 1692 Intron NP_001035869.1
NM_001282792.1 1692 Intron NP_001269721.1
NM_018089.2 1692 Intron NP_060559.2
XM_005246663.2 1692 Intron XP_005246720.1
XM_011511392.2 1692 Intron XP_011509694.1
XM_017004420.1 1692 Intron XP_016859909.1
XM_017004421.1 1692 Intron XP_016859910.1
Gene
ATG9A
Gene Name
autophagy related 9A
There are no transcripts associated with this gene.

Gene
GLB1L
Gene Name
galactosidase beta 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286423.1 1692 Missense Mutation ACC,ATC T635I NP_001273352.1
NM_001286427.1 1692 Missense Mutation ACC,ATC T545I NP_001273356.1
NM_024506.4 1692 Missense Mutation ACC,ATC T635I NP_078782.3
XM_011511813.2 1692 Missense Mutation ACC,ATC T392I XP_011510115.1
XM_017004895.1 1692 Missense Mutation ACC,ATC T545I XP_016860384.1
Gene
STK16
Gene Name
serine/threonine kinase 16
There are no transcripts associated with this gene.

View Full Product Details