Product Details

SNP ID
rs142698865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:216672215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATAGTATACAGCAATCTTCACTT[C/T]TTAAGAAAATGTGAGATCCTTTGTT
Phenotype
MIM: 146731 MIM: 146734
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGFBP2 PubMed Links

Gene Details

Gene
IGFBP2
Gene Name
insulin like growth factor binding protein 2
There are no transcripts associated with this gene.

Gene
IGFBP5
Gene Name
insulin like growth factor binding protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000599.3 6129 UTR 3 NP_000590.1

View Full Product Details