Product Details

SNP ID
rs143333285
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24038260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGACTTTGAAGAAGGAAGAAAT[A/G]TTGTATCAGTGAGTTTTTGTTTTCC
Phenotype
MIM: 600620 MIM: 616234
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FKBP1B PubMed Links

Gene Details

Gene
FKBP1B
Gene Name
FK506 binding protein 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322963.1 1366 Intron NP_001309892.1
NM_001322964.1 1366 Intron NP_001309893.1
NM_004116.4 1366 Intron NP_004107.1
NM_054033.3 1366 Intron NP_473374.1
XM_011532699.2 1366 Intron XP_011531001.1
XM_017003593.1 1366 Intron XP_016859082.1
XM_017003594.1 1366 Intron XP_016859083.1
XM_017003595.1 1366 Intron XP_016859084.1
XM_017003596.1 1366 Intron XP_016859085.1
Gene
WDCP
Gene Name
WD repeat and coiled coil containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142319.1 1366 Missense Mutation ACA,ATA T412I NP_001135791.1
NM_025203.2 1366 Missense Mutation ACA,ATA T412I NP_079479.1
XM_017005029.1 1366 Missense Mutation ACA,ATA T412I XP_016860518.1

View Full Product Details