Product Details

SNP ID
rs143421198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:187465743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAAGCTATTCATGAAGCTAGTC[A/G]TGAAGCTAGTCATACCAAATGAATG
Phenotype
MIM: 152310
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TFPI PubMed Links

Gene Details

Gene
TFPI
Gene Name
tissue factor pathway inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005246818.1 2434 UTR 3 XP_005246875.1
XM_005246819.1 2434 UTR 3 XP_005246876.1
XM_006712720.3 2434 UTR 3 XP_006712783.1
XM_011511707.2 2434 UTR 3 XP_011510009.1
XM_011511708.1 2434 UTR 3 XP_011510010.1
XM_011511709.2 2434 UTR 3 XP_011510011.1

View Full Product Details