Product Details

SNP ID
rs144301087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:164493140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTATTAGATCTGTAAATCTTGTGT[A/G]GCCATCATCCAGTGTGTGGAACATT
Phenotype
MIM: 601524
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRB14 PubMed Links

Gene Details

Gene
GRB14
Gene Name
growth factor receptor bound protein 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303422.1 1497 Missense Mutation CAC,TAC H420Y NP_001290351.1
NM_004490.2 1497 Missense Mutation CAC,TAC H507Y NP_004481.2
XM_011511022.2 1497 Intron XP_011509324.1
XM_017003899.1 1497 Missense Mutation CAC,TAC H307Y XP_016859388.1

View Full Product Details