Product Details

SNP ID
rs144612738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:196766579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCACAATACCTCTACCACAAGTG[C/G]AGGGAGCTCCAGGGCCTTCTGATAA
Phenotype
MIM: 604888
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GTF3C3 PubMed Links

Gene Details

Gene
GTF3C3
Gene Name
general transcription factor IIIC subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206774.1 2681 Intron NP_001193703.1
NM_012086.4 2681 Missense Mutation CCA,GCA P842A NP_036218.1
XM_005246965.4 2681 Missense Mutation CCA,GCA P450A XP_005247022.1

View Full Product Details