Product Details

SNP ID
rs144678998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:178118069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTCTCTTAGGTGTATGGAGATAT[C/T]GAGTATTGCAGCATTATTAAGAATA
Phenotype
MIM: 604961 MIM: 608888
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDE11A PubMed Links

Gene Details

Gene
PDE11A
Gene Name
phosphodiesterase 11A
There are no transcripts associated with this gene.

Gene
RBM45
Gene Name
RNA binding motif protein 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152945.3 1005 Silent Mutation ATC,ATT I146I NP_694453.2
XM_005246287.4 1005 Silent Mutation ATC,ATT I146I XP_005246344.1
XM_017003320.1 1005 Silent Mutation ATC,ATT I289I XP_016858809.1
XM_017003321.1 1005 Silent Mutation ATC,ATT I289I XP_016858810.1
XM_017003322.1 1005 Silent Mutation ATC,ATT I146I XP_016858811.1

View Full Product Details