Product Details

SNP ID
rs145197227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:165090201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTTTGCTTTCTTTTTCTGGTTT[A/G]TCTTTCTCAAACTTTTCCTTGTCTG
Phenotype
MIM: 182391
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCN3A PubMed Links

Gene Details

Gene
SCN3A
Gene Name
sodium voltage-gated channel alpha subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081676.1 6567 Silent Mutation GAC,GAT D1935D NP_001075145.1
NM_001081677.1 6567 Silent Mutation GAC,GAT D1935D NP_001075146.1
NM_006922.3 6567 Silent Mutation GAC,GAT D1984D NP_008853.3
XM_011511610.2 6567 Silent Mutation GAC,GAT D1984D XP_011509912.1
XM_011511613.2 6567 Silent Mutation GAC,GAT D1354D XP_011509915.1
XM_017004660.1 6567 Silent Mutation GAC,GAT D1984D XP_016860149.1
XM_017004661.1 6567 Silent Mutation GAC,GAT D1967D XP_016860150.1
XM_017004662.1 6567 Silent Mutation GAC,GAT D1938D XP_016860151.1
XM_017004663.1 6567 Silent Mutation GAC,GAT D1354D XP_016860152.1
XM_017004664.1 6567 Intron XP_016860153.1
XM_017004665.1 6567 Intron XP_016860154.1
XM_017004666.1 6567 Intron XP_016860155.1

View Full Product Details