Product Details

SNP ID
rs145599521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:178116380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAAGAAGATCTGCGGGAAAAAT[G/T]TAAGGTATTTATTCTAATCAGCTAG
Phenotype
MIM: 604961 MIM: 608888
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PDE11A PubMed Links

Gene Details

Gene
PDE11A
Gene Name
phosphodiesterase 11A
There are no transcripts associated with this gene.

Gene
RBM45
Gene Name
RNA binding motif protein 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152945.3 986 Missense Mutation TGT,TTT C140F NP_694453.2
XM_005246287.4 986 Missense Mutation TGT,TTT C140F XP_005246344.1
XM_017003320.1 986 Missense Mutation TGT,TTT C283F XP_016858809.1
XM_017003321.1 986 Missense Mutation TGT,TTT C283F XP_016858810.1
XM_017003322.1 986 Missense Mutation TGT,TTT C140F XP_016858811.1

View Full Product Details