Product Details

SNP ID
rs145946342
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:237567094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGTCGGAGGAGGGCAACCCTGGG[A/G]GACGTCCCCAAGCCTGGCCTGCGAC
Phenotype
MIM: 602663 MIM: 602206
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRLH PubMed Links

Gene Details

Gene
PRLH
Gene Name
prolactin releasing hormone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015893.1 183 Silent Mutation GGA,GGG G61G NP_056977.1
Gene
RAB17
Gene Name
RAB17, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details