Product Details

SNP ID
rs146200342
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:203870615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACGTGGCCCAGCCTGCTGTGGTA[C/T]TGGCCAGCAGCCGAGGCATCGCCAG
Phenotype
MIM: 123890
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CTLA4 PubMed Links

Gene Details

Gene
CTLA4
Gene Name
cytotoxic T-lymphocyte associated protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037631.2 294 Silent Mutation CTG,TTG L47L NP_001032720.1
NM_005214.4 294 Silent Mutation CTG,TTG L47L NP_005205.2

View Full Product Details