Product Details

SNP ID
rs146588188
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:215942858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTTGAATGTTTATCTTGCGAGA[G/T]TCTACAGAATTGAGAGAAAATATCT
Phenotype
MIM: 609207
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
MREG PubMed Links

Gene Details

Gene
MREG
Gene Name
melanoregulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018000.2 2946 UTR 3 NP_060470.2
XM_011511468.2 2946 UTR 3 XP_011509770.1
XM_017004475.1 2946 Intron XP_016859964.1

View Full Product Details