Product Details

SNP ID
rs146720297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:203706736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGCTCTCAACTTATTCCCTTCA[A/G]TTCAAGTAACAGGTAAACAATGTTA
Phenotype
MIM: 186760
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD28 PubMed Links

Gene Details

Gene
CD28
Gene Name
CD28 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243077.1 262 Missense Mutation ATT,GTT I14V NP_001230006.1
NM_001243078.1 262 Missense Mutation ATT,GTT I14V NP_001230007.1
NM_006139.3 262 Missense Mutation ATT,GTT I14V NP_006130.1
XM_011512194.2 262 Intron XP_011510496.1
XM_011512195.2 262 Intron XP_011510497.1
XM_011512197.2 262 Missense Mutation ATT,GTT I14V XP_011510499.1

View Full Product Details