Product Details

SNP ID
rs147058083
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27384043 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCGGTGTCATCCTCATCTTCC[C/T]CATTCTCTGCCTCCTCACTGCTGTA
Phenotype
MIM: 605119 MIM: 605963 MIM: 613598
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FTH1P3 PubMed Links

Gene Details

Gene
FTH1P3
Gene Name
ferritin heavy chain 1 pseudogene 3
There are no transcripts associated with this gene.

Gene
PPM1G
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177983.2 1136 Missense Mutation GAG,GGG E292G NP_817092.1
Gene
SNX17
Gene Name
sorting nexin 17
There are no transcripts associated with this gene.

Gene
ZNF513
Gene Name
zinc finger protein 513
There are no transcripts associated with this gene.

View Full Product Details