Product Details

SNP ID
rs147183942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:99292556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTACCACAGTAGTTCAGGCCGTGA[C/T]GTCTTCCAATCCCACAAGATGCTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYG1 PubMed Links

Gene Details

Gene
LYG1
Gene Name
lysozyme g1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174898.2 362 Missense Mutation CAT,CGT H43R NP_777558.1
XM_005263869.2 362 Missense Mutation CAT,CGT H43R XP_005263926.1
XM_005263870.3 362 Missense Mutation CAT,CGT H43R XP_005263927.1
XM_005263871.4 362 Missense Mutation CAT,CGT H43R XP_005263928.1
XM_011510580.2 362 Missense Mutation CAT,CGT H43R XP_011508882.1
XM_017003311.1 362 Missense Mutation CAT,CGT H43R XP_016858800.1
XM_017003312.1 362 Missense Mutation CAT,CGT H43R XP_016858801.1
XM_017003313.1 362 Missense Mutation CAT,CGT H43R XP_016858802.1

View Full Product Details