Product Details
- SNP ID
-
rs147318949
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:149575805 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTGTTACAGTCAGAATCATTAGT[G/T]TGCCATTAGCTACTTCTGGAAACAG
- Phenotype
-
MIM: 611935
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MMADHC
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12232959] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MMADHC
- Gene Name
- methylmalonic aciduria and homocystinuria, cblD type
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015702.2 |
|
Intron |
|
|
NP_056517.1 |
View Full Product Details