Product Details

SNP ID
rs147457323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:232540150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCTGGTAAGCCGCAGGACGGAG[A/G]AGGGGTCAGCGCACCACGCCCTGGG
Phenotype
MIM: 100720 MIM: 100730 MIM: 612972
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHRND PubMed Links

Gene Details

Gene
CHRND
Gene Name
cholinergic receptor nicotinic delta subunit
There are no transcripts associated with this gene.

Gene
CHRNG
Gene Name
cholinergic receptor nicotinic gamma subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005199.4 Intron NP_005190.4
Gene
TIGD1
Gene Name
tigger transposable element derived 1
There are no transcripts associated with this gene.

View Full Product Details