Product Details

SNP ID
rs147510562
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:32630673 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAGGTAGTAACATTCCTGGATT[A/T]CTCAACAGATTTGGACACAACGGAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR4765 PubMed Links

Gene Details

Gene
MIR4765
Gene Name
microRNA 4765
There are no transcripts associated with this gene.

Gene
TTC27
Gene Name
tetratricopeptide repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193509.1 512 Missense Mutation TAC,TTC Y30F NP_001180438.1
NM_017735.4 512 Missense Mutation TAC,TTC Y80F NP_060205.3
XM_005264416.1 512 Missense Mutation TAC,TTC Y80F XP_005264473.1
XM_011532958.1 512 Missense Mutation TAC,TTC Y80F XP_011531260.1

View Full Product Details