Product Details

SNP ID
rs147578390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96848368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTCACATGCTAGCCGTGCCTTT[C/T]GGTCCCGGATGGCCTTCAGGGCTGG
Phenotype
MIM: 610736
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD23 PubMed Links

Gene Details

Gene
ANKRD23
Gene Name
ankyrin repeat domain 23
There are no transcripts associated with this gene.

Gene
ANKRD39
Gene Name
ankyrin repeat domain 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016466.5 517 Missense Mutation CAA,CGA Q162R NP_057550.3

View Full Product Details