Product Details

SNP ID
rs148134162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:177616705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAAAGCCACTATCAGTCATAAC[G/T]ACTTATTCCATCCTATAATCTCATA
Phenotype
MIM: 604961
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PDE11A PubMed Links

Gene Details

Gene
PDE11A
Gene Name
phosphodiesterase 11A
There are no transcripts associated with this gene.

Gene
TTC30A
Gene Name
tetratricopeptide repeat domain 30A
There are no transcripts associated with this gene.

View Full Product Details