Product Details

SNP ID
rs149286673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:46618798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAAAAGAAACTTGGTACTGTTAT[C/T]ACTCCAGATACATGGAAAGATGGTG
Phenotype
MIM: 604594 MIM: 600153
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRIPT PubMed Links

Gene Details

Gene
CRIPT
Gene Name
CXXC repeat containing interactor of PDZ3 domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014171.5 153 Silent Mutation ATC,ATT I14I NP_054890.1
Gene
PIGF
Gene Name
phosphatidylinositol glycan anchor biosynthesis class F
There are no transcripts associated with this gene.

View Full Product Details