Product Details

SNP ID
rs149409403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:173910025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGGATATCTTGATTGCTGGTGGC[A/G]GAAGTATTAACAGTTCCTATCCCTG
Phenotype
MIM: 601804
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SP3 PubMed Links

Gene Details

Gene
SP3
Gene Name
Sp3 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017371.4 2784 Silent Mutation TCC,TCT S686S NP_001017371.3
NM_001172712.1 2784 Silent Mutation TCC,TCT S751S NP_001166183.1
NM_003111.4 2784 Silent Mutation TCC,TCT S754S NP_003102.1

View Full Product Details