Product Details

SNP ID
rs149558539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:79137148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGATTAGTCTCATCTCATATCAG[C/T]CACTGAACACACTGGAGAGTGGCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
REG1CP PubMed Links

Gene Details

Gene
REG1CP
Gene Name
regenerating family member 1 gamma, pseudogene
There are no transcripts associated with this gene.

View Full Product Details