Product Details

SNP ID
rs149584805
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:164493133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCAGCTGTATTAGATCTGTAAAT[A/C]TTGTGTGGCCATCATCCAGTGTGTG
Phenotype
MIM: 601524
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
GRB14 PubMed Links

Gene Details

Gene
GRB14
Gene Name
growth factor receptor bound protein 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303422.1 1504 Missense Mutation AGA,ATA R422I NP_001290351.1
NM_004490.2 1504 Missense Mutation AGA,ATA R509I NP_004481.2
XM_011511022.2 1504 Intron XP_011509324.1
XM_017003899.1 1504 Missense Mutation AGA,ATA R309I XP_016859388.1

View Full Product Details