Product Details

SNP ID
rs150014662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70088233 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCGAGTGTGTCAAGCAGATTTGC[C/T]TGGTCATGCTGGAGACGCTCTCCCA
Phenotype
MIM: 601209
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCBP1 PubMed Links

Gene Details

Gene
PCBP1
Gene Name
poly(rC) binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006196.3 781 Silent Mutation CTG,TTG L164L NP_006187.2
Gene
PCBP1-AS1
Gene Name
PCBP1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details