Product Details

SNP ID
rs150482545
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:118962538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTGCCTTTTACTGAAGATTTTT[A/G]TGTCAAAGTTCATGAAATAGTTTTG
Phenotype
MIM: 604870
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MARCO PubMed Links

Gene Details

Gene
MARCO
Gene Name
macrophage receptor with collagenous structure
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006770.3 Intron NP_006761.1
XM_011512082.1 Intron XP_011510384.1
XM_011512083.2 Intron XP_011510385.1
XM_017005171.1 Intron XP_016860660.1

View Full Product Details