Product Details

SNP ID
rs150487874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:29193404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACCTCCTTCATATTGGCAGTCAG[C/T]GAAGAGGGCTCTAGGAGCAGTGAGG
Phenotype
MIM: 105590
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALK PubMed Links

Gene Details

Gene
ALK
Gene Name
anaplastic lymphoma receptor tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004304.4 5635 Silent Mutation TCA,TCG S1561S NP_004295.2
Gene
CLIP4
Gene Name
CAP-Gly domain containing linker protein family member 4
There are no transcripts associated with this gene.

View Full Product Details