Product Details

SNP ID
rs150627833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:196764626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCGGTATTCCCACTGCTCTGATA[C/G]ATGAGAGACAAGTTGTAGGCAATAT
Phenotype
MIM: 604888
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GTF3C3 PubMed Links

Gene Details

Gene
GTF3C3
Gene Name
general transcription factor IIIC subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206774.1 2755 Intron NP_001193703.1
NM_012086.4 2755 Silent Mutation ATC,ATG I866M NP_036218.1
XM_005246965.4 2755 Missense Mutation ATC,ATG I474M XP_005247022.1

View Full Product Details