Product Details
- SNP ID
-
rs150679557
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:182834865 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCGGGATTTAGTTGCGTGCTTGC[C/T]GGGGGTTCGAGTTCCTGCCAGACTT
- Phenotype
-
MIM: 605083
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FRZB
PubMed Links
Gene Details
- Gene
- FRZB
- Gene Name
- frizzled-related protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001463.3 |
1180 |
Missense Mutation |
CAG,CGG |
Q321R |
NP_001454.2 |
View Full Product Details