Product Details

SNP ID
rs150809496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:232540688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGCCGTCCACCATGGTGTGGCG[A/G]CCGGATATCGTGCTGGAGAACAAGT
Phenotype
MIM: 100720 MIM: 100730 MIM: 605895 MIM: 612972
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHRND PubMed Links

Gene Details

Gene
CHRND
Gene Name
cholinergic receptor nicotinic delta subunit
There are no transcripts associated with this gene.

Gene
CHRNG
Gene Name
cholinergic receptor nicotinic gamma subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005199.4 348 Silent Mutation CGA,CGG R109R NP_005190.4
Gene
EIF4E2
Gene Name
eukaryotic translation initiation factor 4E family member 2
There are no transcripts associated with this gene.

Gene
MIR5001
Gene Name
microRNA 5001
There are no transcripts associated with this gene.

Gene
TIGD1
Gene Name
tigger transposable element derived 1
There are no transcripts associated with this gene.

View Full Product Details